CAS RN 1026936-07-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac Ketorolac 6-Benzoyl Isomer, TRC
CAS: 1026936-07-1 Molecular Formula: C15 H13 N O3 Molecular Weight (g/mol): 255.27 Synonym: 6-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic Acid,(1RS)-6-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic Acid IUPAC Name: 6-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid SMILES: OC(=O)C1CCn2cc(cc12)C(=O)c3ccccc3