accessibility menu, dialog, popup

UserName

CH3OHNHOO

CAS RN 10242-01-0

IUPAC Name: 5-methoxy-1H-indole-3-carboxylic acid
Synonyms: 5-methoxy-1H-indole-3-carboxylic acid
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: RVVSEZGJCOAUED-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC=C(C(O)=O)C2=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results