accessibility menu, dialog, popup

UserName

CAS RN 1017779-90-6

CH3OFOClF

CAS RN 1017779-90-6

IUPAC Name: 3-ethoxy-2,4-difluorobenzoyl chloride
Synonyms: 3-ethoxy-2,4-difluorobenzoyl chloride
Molecular Weight (g/mol): 220.6
Molecular Formula: C9H7ClF2O2
InChi Key: WMLHUPZOTFCPNP-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C=CC(C(Cl)=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results