accessibility menu, dialog, popup

UserName

CH3OFOHF

CAS RN 1017779-35-9

IUPAC Name: (4-ethoxy-3,5-difluorophenyl)methanol
Synonyms: (4-ethoxy-3,5-difluorophenyl)methanol
Molecular Weight (g/mol): 188.17
Molecular Formula: C9H10F2O2
InChi Key: TUTXCECYJMMZLA-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C=C(CO)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results