accessibility menu, dialog, popup

UserName

H3CH3CSOONOHOCH3CH3

CAS RN 1009282-06-7

IUPAC Name: 1-(2,3,5,6-tetramethylbenzenesulfonyl)pyrrolidine-2-carboxylic acid
Synonyms: 1-(2,3,5,6-tetramethylbenzenesulfonyl)pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 311.4
Molecular Formula: C15H21NO4S
InChi Key: DFFZRBIPGBTBHG-UHFFFAOYNA-N
SMILES: CC1=CC(C)=C(C)C(=C1C)S(=O)(=O)N1CCCC1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)