Learn More
Greiner Bio-One MASTERBLOCK™ 96 Deep Well Round Bottom 1mL Storage Plate
Suitable for cultivating bacteria or yeasts. Greiner Bio-One MASTERBLOCK™ 96 Deep Well Round Bottom 1mL Storage Plate is also suitable for storing sample material.
418.00€
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Material | Polypropylene |
No. of Wells | 96 |
Total Volume (Metric) Well | 1 mL |
Product Code | Brand | Sterility | No. per Pack | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | No. per Pack | Price | Quantity & Availability | ||||
10013381
|
Greiner Bio-One
780261 |
Sterile | 1 |
418.00€
Pack of 50 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Sealable with adhesive films and heat sealer
- High temperature resistance
- High chemical resistance
- A
Specifications
Standard | |
Polypropylene | |
1 mL | |
50/Cs. |
8 x 12 | |
96 | |
Round Bottom (U-well) | |
Chimney |