Learn More
Cole-Parmer™ Digi-Sense™ USB Temperature and Temperature/RH Dataloggers
Product Code. 15890006
Click to view available options
Display Type:
LCD
No Display
Status Indicator Lights
Humidity Range:
0 to 100%
No humidity monitoring
Temperature Range (Metric):
-200°C to +1370°C
-40°C to +70°C
Unit Size:
Each
Product Code. 15890006
Brand: Cole-Parmer™ 2025044
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Digi-Sense™ USB Dataloggers are portable or mountable to fit multiple applications
  • Each Digi-Sense data logger comes user ready with software, mounting clip and easy to following instructions.
  • Intended applications include monitoring pharmaceutical, food & beverage, storage and office, museum, laboratory and transportation environments.
  • Ease of use and affordability make these instruments ideal for any need where accurate temperature monitoring is needed.
  • Intuitive software supports two modes of data display and powerful data scaling and panning functionality. Data can be downloaded in spreadsheet compatible formats for analysis or graphically printed out.
  • The durable polycarbonate housing and precision sensors and electronics are backed by a 2-year warranty.
Warranty
2 Years
Specifications
Type Datalogger
Display Type Status Indicator Lights
Data Memory 32,000 Readings
Calibration Not Calibrated
Alarm Types High/Low, Selectable
Software Type Setup, real-time monitoring, data acquisition, data analysis
Battery Type 3.6 V Lithium, 1/2 AA
Battery Life 1 Year Typical
Includes Software, mounting clip and hardware, protective plastic cover, and battery.
Channels 1
Interface USB
Material Plastic
For Use With (Application) Monitoring pharmaceutical, food & beverage, storage and office, museum, laboratory and transportation environments. 
For Use With (Equipment) Type-K Thermocouple Probes
Accuracy (Temperature) ±2°F (1°C)
Humidity Range No humidity monitoring
Temperature Resolution 0.01°
Temperature Range (Metric) -200°C to +1370°C
Length (English) 3.875 in.
Length (Metric) 9.8 cm
Weight (Metric) 0.4 kg