Learn More
Macherey-Nagel™ Chromabond™ Florisil Glass SPE Column
Click to view available options
Bed Weight:
1 g
2 g
Unit Size:
Pack of 30
Pack of 4
Specifications
Specifications
| Base Material | Magnesium Silicate |
| Column Capacity (Metric) | 6 mL |
| Column Type | SPE |
| Bed Weight | 2 g |
| Particle Size | 150 to 250 μm |
| Sorbent | Chromabond Florisil |
| Type | Solid Phase Extraction Column |