Learn More
Equilenin-4,16,16-d3, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Equilenin-d3 |
| CAS | 56588-54-6 |
| Molecular Formula | C18 2H3 H15 O2 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C18H18O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h2-5,10,16,19 H,6-9H2,1H3/t16-,18-/m0/s1/i7D2,10 D |
| SMILES | [2 H]c1c(O)ccc2c3CC[C@@]4(C)[C@@H](CC([2 H])([2 H])C4=O)c3ccc12 |
| IUPAC Name | (13 S,14 S)-4,16,16-trideuterio-3-hydroxy-13-methyl-11,12,14,15-tetrahydrocyclopenta[a]phenanthren-17-one |
| Molecular Weight (g/mol) | 269.35 |
| Formula Weight | 269.1495 g/mol |
| Percent Purity | 97 atom % D, min 98% Chemical Purity |