missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc chloride, Free-flowing, Reagent Grade, anhydrous, ≥98%, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | 293°C (literature) |
CAS Max % | 100.0000% |
Physical Form | Anhydrous |
Quantity | 1 kg |
Linear Formula | ZnCl2 |
UN Number | UN2331 |
Formula Weight | 136.3 |
Percent Purity | >97% |
Grade | ACS Reagent |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'