missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride, ≥97%, Honeywell™ Fluka™

Catalog Number p-7120912
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
7646-85-7
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
This item is not returnable. View return policy
7646-85-7
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl

Free-flowing, ACS reagent, anhydrous,≥97%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 287°C to 304°C
Color White
pH 5 (at 20°C)
Physical Form Solid
Quantity 100 g
Linear Formula ZnCl2
UN Number UN2331
Solubility Information 851g/L
Formula Weight 136.3g/mol
Percent Purity ≥97%
Odor Odorless
Packaging Poly bottle
Vapor Pressure 1mmHg (428°C)
Density 2.93g/cm3
Chemical Name or Material Zinc chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'