missing translation for 'onlineSavingsMsg'
Learn More

Binder™ Zero Voltage Alarm Output

Binder™ Zero Voltage Alarm Output for temperature and humidity values.

Brand:  Binder™ 8012-1001

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15622046

  • 807.00€ / Each


please call +351214253350
Add to basket

Description

Description

  • Temperature ±2°C, humidity ±5% RH
  • Accessible via 6-pin DIN socket (max. 24 V – 2.5 A)
  • With audible signal that can be switched off
Specifications

Specifications

KT 115
Temperature and humidity
Alarm Output
Videos
Documents