Learn More
Abnova™ YBX1/CEN1p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
1475.00€ - 2430.00€
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | Y box binding protein 1 |
Gene Alias | BP-8,CSDA2,CSDB,DBPB,MDR-NF1,MGC104858,MGC110976,MGC117250,NSEP-1,NSEP1,YB-1,YB1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16033365
|
Abnova™
FG0223.100UL |
100 μL |
1475.00€
100µL |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
16043365
|
Abnova™
FG0223.200UL |
200 μL |
2430.00€
200µL |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
BP-8,CSDA2,CSDB,DBPB,MDR-NF1,MGC104858,MGC110976,MGC117250,NSEP-1,NSEP1,YB-1,YB1 | |
4904 | |
Liquid |
Tube | |
Y box binding protein 1 | |
YBX1 | |
DNA/RNA |