missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Xq27/CENXp FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
1475.00€ - 2430.00€
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Product Type | DNA/RNA |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013315
|
Abnova™
FG0196.100UL |
100 μL |
1475.00€
100µL |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
16023315
|
Abnova™
FG0196.200UL |
200 μL |
2430.00€
200µL |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
Liquid |
Tube | |
DNA/RNA |