Learn More
Eppendorf™ Xplorer™ Plus, Multi-Channel, Variable
The future in your hands. With the Eppendorf Xplorer/Xplorer plus electronic pipette, your work achieves a new level of simplicity, precision and reproducibility.
1341.00€ - 1483.00€
Specifications
Adjustable Tip Spacing No
Autoclavable yes (lower part)
Battery Type Li-Ion
Finger Style Ergonomic
Product Type Electronic Pipette
View More Specs
Products 4
Product Code Brand Volume (Metric) Number of Channels Price Quantity & Availability  
Product Code Brand Volume (Metric) Number of Channels Price Quantity & Availability  
16262401 View Documents Eppendorf™
4861000778
20 µL to 1 µL 16 Channels 1341.00€
Each View alternative products
Save Up to 
»
Estimated Shipment
24-09-2019
Add to basket
16282401 View Documents Eppendorf™
4861000792
100 µL to 5 µL 16 Channels 1341.00€
Each View alternative products
Save Up to 
»
Estimated Shipment
24-09-2019
Add to basket
16272401 View Documents Eppendorf™
4861000779
20 µL to 1 µL 24 Channels 1483.00€
Each View alternative products
Save Up to 
»
Estimated Shipment
24-09-2019
Add to basket
16292401 View Documents Eppendorf™
4861000793
100 µL to 5 µL 24 Channels 1483.00€
Each View alternative products
Save Up to 
»
Estimated Shipment
24-09-2019
Add to basket
Description
Description
  • Multi-functional rocker with up is up and down is down functionality
  • Selection dial for quick and easy selection of functions
  • Self-explanatory color display without submenus and with ergonomic angle for optimal readability in every position
  • Spring-loaded tip cone for all pipettes up to 1,000 µL for minimal tip attachment forces helps to reduce stress
  • Powerful rechargeable battery for up to 8 hours work without the need for recharging
  • Separate power socket: Work can continue while charging
  • Adjustment to external conditions (e.g. geographic height, liquids with high density)
  • Lower part of pipette can be autoclaved to ensure decontamination
  • 9 different languages: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish
  • Xplorer plus only: Individual programming, fixed volume settings, password protection, on/off switch, additional applications, display of service intervals
Specifications
Specifications
No
Li-Ion
Electronic Pipette
yes (lower part)
Ergonomic