Learn More
Eppendorf™ Xplorer™ Plus, Multi-Channel, Variable
The future in your hands. With the Eppendorf Xplorer/Xplorer plus electronic pipette, your work achieves a new level of simplicity, precision and reproducibility.
1341.00€ - 1483.00€
Specifications
| Adjustable Tip Spacing | No |
|---|---|
| Autoclavable | yes (lower part) |
| Battery Type | Li-Ion |
| Finger Style | Ergonomic |
| Product Type | Electronic Pipette |
| Product Code | Brand | Volume (Metric) | Number of Channels | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Volume (Metric) | Number of Channels | Price | Quantity & Availability | ||||
| 16262401 |
Eppendorf™
4861000778 |
20 µL to 1 µL | 16 Channels |
1341.00€ Each |
|
16282401 |
Eppendorf™
4861000792 |
100 µL to 5 µL | 16 Channels |
1341.00€ Each |
|
16272401 |
Eppendorf™
4861000779 |
20 µL to 1 µL | 24 Channels |
1483.00€ Each |
|
16292401 |
Eppendorf™
4861000793 |
100 µL to 5 µL | 24 Channels |
1483.00€ Each |
|
Description
- Multi-functional rocker with up is up and down is down functionality
- Selection dial for quick and easy selection of functions
- Self-explanatory color display without submenus and with ergonomic angle for optimal readability in every position
- Spring-loaded tip cone for all pipettes up to 1,000 µL for minimal tip attachment forces helps to reduce stress
- Powerful rechargeable battery for up to 8 hours work without the need for recharging
- Separate power socket: Work can continue while charging
- Adjustment to external conditions (e.g. geographic height, liquids with high density)
- Lower part of pipette can be autoclaved to ensure decontamination
- 9 different languages: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish
- Xplorer plus only: Individual programming, fixed volume settings, password protection, on/off switch, additional applications, display of service intervals
Specifications
| No | |
| Li-Ion | |
| Electronic Pipette |
| yes (lower part) | |
| Ergonomic |