Learn More
Cryo-Industrial™ Wrist Fabric Gloves
Cryo-Industrial™ Wrist Fabric Gloves provide protection for hands and arms when working in hazardous, ultra-cold environments down to -200°C
Brand:  Cryo-Industrial™ CIWLWP
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11873851
Quantity Price
1 196.00€ / 1 pair
6 169.60€ / 1 pair
Estimated Shipment
15-03-2021
Add to basket
Description
Description
  • Protection for hands and arms when working in hazardous, ultra-cold environments down to -200°C
  • Gloves are designed to provide a high level of thermal protection and abrasion resistance for heavy-duty applications
  • Wrist length
  • Made from state-of-the-art materials
  • Designed to keep the user dry and to wick moisture away from your hands
  • Can be worn in comfort for extended periods of time.
  • Multi-layered construction allows for a maximum level of thermal protection, flexibility, and dexterity
Specifications
Specifications
No
Fabric
Powder-free
Non-sterile
Wrist-length
Large