Learn More
Cryo-Industrial™ Wrist Fabric Gloves
Click to view available options
Size:
Large
X-Large
Description
- Protection for hands and arms when working in hazardous, ultra-cold environments down to -200°C
- Gloves are designed to provide a high level of thermal protection and abrasion resistance for heavy-duty applications
- Wrist length
- Made from state-of-the-art materials
- Designed to keep the user dry and to wick moisture away from your hands
- Can be worn in comfort for extended periods of time.
- Multi-layered construction allows for a maximum level of thermal protection, flexibility, and dexterity
Specifications
Color | Blue |
Disposable | Reusable |
Material | Fabric |
Powdered | Powder Free |
Product Type | Cryogenic Gloves |
Size | X-Large |
Sterility | Non-sterile |
Style | Wrist-length |