Learn More
Cytiva Whatman™ Quartz Air Sampling Filters, Grade QM-A, Circle
Used in many air sampling filter applications, including sampling of acidic gases, stacks, flues, and aerosols. Whatman™ Quartz Air Sampling Filters are sequentially numbered according to U.S. Environmental Protection Agency (EPA) standards and are suitable for most applications.
244.00€ - 875.00€
Specifications
Format Circle
Grade Grade QM-A
View More Specs
Products 13
Product Code Brand Diameter (Metric) Filter Price Quantity & Availability  
Product Code Brand Diameter (Metric) Filter Price Quantity & Availability  
11330624
View Documents Promotion Details
Cytiva
1851-025
25 mm
244.00€
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Add to basket
11303345
View Documents Promotion Details
Cytiva
1851-101
101.6 mm
530.00€
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
Add to basket
11313355
View Documents Promotion Details
Cytiva
1851-085
85 mm
585.00€
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 22-04-2024
to see stock.
Add to basket
11393345
View Documents Promotion Details
Cytiva
1851-050
50 mm
356.00€
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
Add to basket
11343345
View Documents Promotion Details
Cytiva
1851-082
82 mm
451.00€
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
Add to basket
11383345
View Documents Promotion Details
Cytiva
1851-150
150 mm
875.00€
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
Add to basket
11353345
View Documents Promotion Details
Cytiva
1851-090
90 mm
595.00€
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
Add to basket
11393335
View Documents Promotion Details
Cytiva
1851-045
45 mm
310.00€
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 30-08-2024
to see stock.
Add to basket
11373345
View Documents Promotion Details
Cytiva
1851-118
118 mm
760.00€
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 30-08-2024
to see stock.
Add to basket
11313345
View Documents Promotion Details
Cytiva
1851-032
32 mm
269.00€
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 22-04-2024
to see stock.
Zum Warenkorb hinzufügen
11340624
missing translation for 'viewDocuments' Promotion Details
Cytiva
1851-037
37 mm
279.00€
Pack of 100
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
In Stock
to see stock.
Zum Warenkorb hinzufügen
10455211
missing translation for 'viewDocuments' Promotion Details
Cytiva
1851-047
47 mm
318.00€
Pack of 100
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 18-04-2024
to see stock.
Zum Warenkorb hinzufügen
11323345
missing translation for 'viewDocuments' Promotion Details
Cytiva
1851-055
55 mm
365.00€
Pack of 100
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-08-2024
to see stock.
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Used in many air sampling filter applications, including sampling of acidic gases, stacks, flues, and aerosols. Whatman™ Quartz Air Sampling Filters are sequentially numbered according to U.S. Environmental Protection Agency (EPA) standards and are suitable for most applications.
  • Temperature resistant up to 500°C
  • Binder-free
  • High loading capacity
  • Fast flow rates (air flow rate 6.4 s/100 ml/in2)
  • Used in many air sampling filter applications, including sampling of acidic gases, stacks, flues, and aerosols
  • Filters are suitable for PM-10 sampling and analysis
Spezifikation
Spezifikation
Circle
Grade QM-A
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate