Learn More
Cytiva Whatman™ Baby Food Testing Filter Papers
Filter mat made from a mixture of cellulose and polyester used for optically testing baby food (artificial milk) for textile fibers.
Brand:  Cytiva 10348903
Additional Details : Weight : 0.23000kg
Product Code. 11378784
  • 281.00€ / Pack of 1000
Estimated Shipment: 18-04-2024
Description
Filter mat made from a mixture of cellulose and polyester used for optically testing baby food (artificial milk) for textile fibers.
Whatman offers a line of cellulose filter papers for specific applications. The product line includes filter papers for use in soil analysis and filter papers for the sugar industry.
Specifications
Specifications
Disc
Optically testing baby food (artificial milk) for textile fibers
0.86 mm
1.25 in.
Cellulose/Polyester
0048
Baby Food Testing Filter Paper
32 mm