Learn More
Zwiebel™ Weights Set
Manufactured to the highest standard. Zwiebel™ Weights Set are precise measuring tools.
Brand: Zwiebel™ PBI56853
Additional Details : Weight : 0.80000kg
Description
- Stainless steel calibration weight
- Delivered with a calibration certificate guaranteeing traceability/In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Calibration Weight | |
Antimagnetic Stainless Steel | |
6110 g total | |
Case includes gripper |
Cofrac calibration certificate | |
Polypropylene | |
Cylindrical |