Learn More
BINDER™ Water Cooling
For Use With: BINDER units MKF 115
Brand:  BINDER™ 8012-0843
Additional Details : Weight : 1.00000kg
Product Code. 15621936
  • 3862.00€ / Each
Estimated Shipment: 18-06-2024
Description
  • For use with climate chambers
  • Available for a variety of models
Specifications
Specifications
Water cooling - for reduced heat loss to ambient air
BINDER Chambers MKF 115
Water Cooling
Reducing Heat Loss