Learn More
Uvex™ Waist Jacket 8865-Anthracite
Catalog Number 17206501
Click to view available options
Size:
106/110
40/42
44/46
48/50
52/54
56/58
60/62
64/66
90/94
98/102
These work clothes were optimised with clothing physiology in mind, and stand out thanks to their sporty lines and ergonomically placed stretch inserts. This makes for excellent wearer comfort and mobility. The design is based on our uvex protection perfect protective clothing collection - this lets you ensure that your employees dress uniformly, even for very different activities.
  • Color: anthracite
  • Material: 65% polyester, 35% cotton ca. 250 g/m², twill weave
  • This is an optimized jacket on physiological points of view with sporty alignment of style and with ergonomically placed stretch insert.
  • Contrasting colored inserts are the optical highlight.
  • Ergonomically shaped, preformed elbow for perfect fit, large stretch zones in the shoulder and arm area for a great extent on ease of movement, many different pockets do offer enough storage space, width-adjustable seam and cuffed sleeves with press studies, extended back part for more safety in kidney and loins areas, suitable for industrial laundering
  • Öko-Tex Standard 100
  • Sizes: 40/42 - 64/66, 90/94 - 106/110
  • Care instructions: 60°C washable, chlorinate impossible, ironing: II points, dry cleaning: P, tumble dryer: I point, suitable for industrial laundering
Size 98/102
Gender Men's
Color Gray
Closure Type Zipper
Disposable No
Thickness 245 g/m²