Learn More
VistaLab™ Technologies MLA™ Brand Micro D-Tipper™ Pipetters
Color coded body for easy volume identification and individually serialized with Calibration Certificate included.
322.00€
Specifications
Compatible Tips | Standard MLA small tip |
---|---|
For Use With | Standard MLA Small Tip |
Material | Aluminum, Stainless Steel |
Model | MLA Micro D-Tipper™ |
Number of Channels | 1 |
Product Code | Brand | Color | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Volume (Metric) | Price | Quantity & Availability | ||||
11975912
|
VistaLab™ Technologies
1802 |
Red | 2 μL |
322.00€
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
11995912
|
VistaLab™ Technologies
1805 |
Silver | 5 μL |
322.00€
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
11945922
|
VistaLab™ Technologies
1810 |
Orange | 10 μL |
322.00€
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
11965912
|
VistaLab™ Technologies
1803 |
Gray | 3 μL |
322.00€
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
11985912
|
VistaLab™ Technologies
1804 |
White | 4 μL |
322.00€
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
11905922
|
VistaLab™ Technologies
1806 |
Green | 6 μL |
322.00€
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
11915922
|
VistaLab™ Technologies
1807 |
Purple | 7 μL |
322.00€
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
11925922
|
VistaLab™ Technologies
1808 |
Black | 8 μL |
322.00€
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
11935922
|
VistaLab™ Technologies
1809 |
Blue | 9 μL |
322.00€
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
Description
- Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts)
- Two-stroke 'To deliver' operation
- Built in tip-ejector
- Glove guard
- Light plunger forces
Specifications
Standard MLA small tip | |
Aluminum, Stainless Steel | |
1 | |
1 Ea. | |
9 in. | |
1.5 in. |
Standard MLA Small Tip | |
MLA Micro D-Tipper™ | |
Pipette | |
Limited Lifetime | |
2 in. |