missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies Pipette Tips: Low Retention
For use with Ovation and MLA Brand pipetters
Brand: VistaLab™ Technologies 4070-1332LR
Additional Details : Weight : 0.00100kg
Description
- Sterile, Low Retention tips packaged in VistaRak: 5 racks of 192 tips each
- Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, etc.
- Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
- All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time
Specifications
25 µL | |
Sterile | |
Blue | |
Not Autoclavable | |
Polypropylene, Polyethylene filters | |
5 in. | |
2 μL Ovation Pipettes Only | |
Ovation™ M | |
Low Retention | |
192 Tips/Rack |
Filtered | |
Clear | |
No | |
Straight | |
Yes | |
Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween, PCR or other biological applications | |
e-beam | |
Rack | |
VistaRack: 5 racks |