Learn More
Macherey-Nagel™ Visocolor™ HE Ammonium High Sensitive Titration Kit
Product Code. 10015220 Shop All Macherey-Nagel Products
Change view
Click to view available options
Range:
0 to 0.50mg/L NH4
Unit Size:
1 test
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
10015220 0 to 0.50mg/L NH4 1 test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10015220 Supplier Macherey-Nagel™ Supplier No. 920006
 Restricted Item
In Stock
Specifications
Quantity 1 Each
Range 0 to 0.50mg/L NH4
Sufficient For 110 Determinations
Type HE Ammonium High Sensitive Titration Kit
compliance-icons
Product Identifier
  • VISOCOLOUR HE ammonium
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide, tri-sodium citrate, sodium nitroprusside,sodium saliylate