missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Viomycin

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Viomycin is a member of the tuberactinomycin family of antibiotics. Inhibits group I intron splicing and prokaryotic protein synthesis. Freezes bacterial ribosomes in either the pre- or post-translational state. Facilitates trans-cleavage of the Neurospora VS ribozyme.
Specifications
Specifications
| CAS | 32988-50-4 |
| Quantity | 10 mg |
| Molecular Formula | C25H43N13O10 |
| Formula Weight | 881.85 |
| Product Type | Viomycin |
| Purity | >98% |
| Molecular Weight (g/mol) | 685.7 |
| Chemical Name or Material | Viomycin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction