Learn More
VE-cadherin Monoclonal Antibody (BV9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1198
Additional Details : Weight : 0.01000kg
Description
MA1-198 VE-Cadherin antibody detects human VE-cadherin by western blot, immunofluorescence and flow cytometry. This monoclonal antibody recognizes the extracellular domain of the VE-cadherin protein which is highly expressed at intercellular junctions of vascular endothelial cells such as Huvec.
VE-cadherin Monoclonal antibody specifically detects VE-cadherin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
VE-cadherin | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P33151 | |
CDH5 | |
Recombinant protein corresponding to the extracellular domain of human VE-Cadherin. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Western Blot | |
BV9 | |
Unconjugated | |
CDH5 | |
7B4; 7B4 antigen; 7B4/cadherin-5; AA408225; cadherin 5; cadherin 5, type 2 (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular epithelium); cadherin-5; cadherin-5; cd144 antigen; VE cadherin; Cd144; cd144 antigen; CDH5; cell-cell adhesion protein; endothelial-specific cadherin; vascular endothelial cadherin; vascular endothelial cadherin precursor; Vec; VEcad; VE-Cad; VE-cadherin; VECD | |
Mouse | |
Protein G | |
RUO | |
1003 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |