Learn More
Cole-Parmer™ VapLock™ Waste Funnels
VapLock™ Waste Funnels safely and securely capture waste and hazardous solvents
Brand:  Cole-Parmer™ SF-005
Additional Details : Weight : 0.91000kg
 View more versions of this product
Product Code. 15824534
Quantity Price
1 528.37€ / Each
5 439.02€ / Each
Estimated Shipment: 22-10-2021
Add to basket
Description
Description
  • Reduces fumes and harmful vapor exhaust
  • Easily connect to Justrite™ Centura safety containers
Services
None
Specifications
Specifications
Solvent Safety
Justrite Centura Containers
SS and PTFE quick disconnect, HDPE funnel
Waste Funnel, 4 in. dia