Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys
VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys connect to waste containers for a secure closed system
Brand:  Cole-Parmer™ WM63X-10
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 15814624
Quantity Price
1 561.46€ / Each
5 466.06€ / Each
Estimated Shipment: 19-10-2021
Add to basket
Description
Description
  • Select from a variety of cap sizes to fit your container
  • Different materials, numbers and sizes of ports help customize to your application
Services
None
Specifications
Specifications
Solvent Safety
PTFE
1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
Solvent-Compatible Containers
63 mm
Solvent Waste Manifold, PTFE, 63 mm, 1 x 1/2 inNPTF, 3 x 1/4 inNPTF, 6 x 1/4 in.-28 UNFF; 1/ea