Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys
VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys connect to waste containers for a secure closed system
Brand:  Cole-Parmer™ WM60P-4
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 15824614
Quantity Price
1 339.59€ / Each
6 282.37€ / Each
Estimated Shipment: 19-10-2021
Add to basket
Description
Description
  • Select from a variety of cap sizes to fit your container
  • Different materials, numbers and sizes of ports help customize to your application
Services
None
Specifications
Specifications
Solvent Safety
Polypropylene
1/2 in. NPT(F), 1/4 in. NPT(F)
Solvent-Compatible Containers
60 mm
Solvent Waste Manifold, PP, 60 mm, 1 x 1/2 in. NPTF, 3 x 1/4 in. NPTF; 1/ea