Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Bottles and Carboys
VapLock™ Solvent Waste Kits for Bottles and Carboys collect effluent from multiple sources while minimizing harmful solvent exhaust
Brand:  Cole-Parmer™ WAK60P-10
Additional Details : Weight : 0.91000kg
 View more versions of this product
Product Code. 15824554
  • 411.53€ / Each
Estimated Shipment: 22-10-2021
Add to basket
Description
Description
Various cap sizes available to fit your solvent container
Services
None
Specifications
Specifications
Solvent Safety
Polypropylene
1 x 1/2 in. NPT(F), 3 x 1/4 in. NPT(F), 6 x 1/4 in.-28 UN(F)(F)
Solvent-Compatible Containers
DIN 60
Solvent Waste Kit