Learn More
Cole-Parmer™ VapLock™ Solvent Delivery Caps with Inlet Valve and Filter
VapLock™ Solvent Delivery Caps with Inlet Valve and Filter reduce chemical evaporation and exposure to hazardous vapors
Brand:  Cole-Parmer™ S38NXFV-2D
Additional Details : Weight : 0.11000kg
 View more versions of this product
Product Code. 15814494
Quantity Price
1 106.50€ / Each
10 88.55€ / Each
Estimated Shipment: 19-10-2021
Add to basket
Description
Description
Use with semi-rigid tubing equal to or less than 1/8 in (3.2 mm) OD
Services
None
Specifications
Specifications
Solvent Safety
White PTFE, PP, SS, FFKM
1/4 in.-28 UNF(F)
Solvent-Compatible Containers
Nalgene 38-430
Two 1/4 in.-28 ports