Learn More
Cole-Parmer™ VapLock™ Adapters for Exhaust Filters, Quick-Disconnect
VapLock™ Adapters for Exhaust Filters, Quick-Disconnect feature various adapter configurations allowing quick and easy connection to emission sources
Brand:  Cole-Parmer™ FA-032
Additional Details : Weight : 0.11000kg
 View more versions of this product
Product Code. 15814464
Quantity Price
1 185.09€ / Each
6 153.23€ / Each
Estimated Shipment: 25-10-2021
Add to basket
Description
Description
  • Fit VapLock exhaust filters, which have 1/4 in NPT(F) ports
  • Features Justrite Centura quick disconnect 
Services
None
Specifications
Specifications
Solvent Safety
Gray/Black SS/PTFE/Acetal
1/4 in. NPT(M)
VapLock Exhaust Filters
NPT(M) x Quick Disconnect, adjustable
Adjustable Filter Adapter, Justrite Centura; 1/ea