Learn More
Cole-Parmer™ VapLock™ Adapters for Exhaust Filters, NPT(M)
VapLock™ Adapters for Exhaust Filters, NPT(M) feature various adapter configurations allowing quick and easy connection to emission sources
Brand:  Cole-Parmer™ FA-014
Additional Details : Weight : 0.11000kg
 View more versions of this product
Product Code. 15814454
Quantity Price
1 20.90€ / Each
10 17.40€ / Each
Estimated Shipment: 25-10-2021
Add to basket
Description
Description
  • Fit VapLock exhaust filters, which have 1/4-inch NPT(F) ports
  • May require additional adapters to fit your container
Services
None
Specifications
Specifications
Solvent Safety
Gray Polypropylene
1/2 in. NPT(M) x 1/4 in. NPT(M)
VapLock Exhaust Filters
90 elbow, 1/4 in. NPT(M) x 1/2 in. NPT(M)
Filter Adapter; 1/ea