Learn More
Cole-Parmer™ VapLock™ Activated Media Exhaust Filters
VapLock™ Activated Media Exhaust Filters reduce harmful chemical emissions and odors from solvent containers
Brand:  Cole-Parmer™ EF-100-102
Additional Details : Weight : 0.23000kg
 View more versions of this product
Product Code. 15824444
Quantity Price
1 148.90€ / Each
10 133.89€ / Each
Estimated Shipment: 25-10-2021
Add to basket
Description
Description
Use with indicators to set a replacement schedule
Services
None
Specifications
Specifications
Solvent Safety
Activated Carbon with Breakthrough Indicator
Exhaust Filter with Indicator, 75 g activated carbon, 1/4 in. NPT(F); 1/ea
VapLock Solvent Containment Systems
1/4 in. NPT(F)