missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vα2 TCR Rat anti-Mouse, FITC, Clone: B20.1, BD
Rat Monoclonal Antibody
145.00€ - 356.00€
Specifications
Antigen | Vα2 TCR |
---|---|
Clone | B20.1 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15881629
|
BD Biosciences
562085 |
25 μg |
145.00€
Each |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
15827128
|
BD Biosciences
553288 |
0.25 mg |
356.00€
Each |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
Description
The B20.1 monoclonal antibody specifically binds to most members of the Vα2 T-cell Receptor (TCR) subfamily in mice having the a, b, and c haplotypes of the Tcrb α haplotypes. Moreover, the frequencies of Vα2+ CD8+ and CD4+ T cells are influenced by H-2 haplotypes.
Specifications
Vα2 TCR | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Immunology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a λ | |
Affinity Purified |
B20.1 | |
Flow Cytometry | |
FITC | |
RUO | |
Mouse | |
Soluble αβ TCR from mouse cytotoxic T-cell clone KB5-C20 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.