Learn More
Tocris Bioscience™ uPSEM 817 tartrate
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
uPSEM 817 tartrate is a selective ultrapotent PSEM (uPSEM) agonist for α7L131G,Q139L,Y217F-GlyR (PSAM4-GlyR) and PSAM4-5-HT3 chimeric ion channels (EC50 values are 0.3 and 0.5 nM, respectively). Suppresses firing of layer 2/3 cortical neurons expressing PSAM4-GlyR in brain slices. Increases contralateral rotation in mice expressing PSAM4-GlyR unilaterally in the substantia nigra reticulata (LED 0.1 mg/kg).
Specifications
Specifications
| Chemical Name or Material | 2-Propoxy-7,8,9,10-tetrahydro-6 H-6,10-methanoazepino[4,5-g]quinoxaline L-tartrate |
| CAS | 2341833-14-3 |
| Quantity | 25 mg |
| Target | Pharmacologically Selective Effector Molecules/PSEM |
| Molecular Formula | C16H19N3O.C4H6O6 |
| Formula Weight | 419.43 |
| Purity | 98% |