Learn More
Greiner Bio-One Universal Sterile Filter Tips With Graduation
Prevent contaminations with liquids and aerosols during pipetting process.
Brand: Greiner Bio-One 739288
Additional Details : Weight : 1.33000kg
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Non-cytotoxic
- Prevents aspiration of particles into interior of pipette
- Minimizes cross-contamination
- Ultra-micro filter made of hydrophobic polyethyl
Specifications
200 μL | |
Sterile | |
Not Autoclavable | |
Irradiation | |
Yes |
Filtered | |
Natural | |
−20° to 80°C | |
Anthos™ New Line, Costar™ Onepette™, Eppendorf™ Research™, Thermo Scientific™ Finnpipette™ Digital, Gilson™ Pipetman™, Gilson™ Pipetman™ P200, Human™ Pipettes | |
Polypropylene |