Learn More
Greiner Bio-One Universal Sterile Filter Tips for Gilson™ P1000
Prevents contamination with liquids and aerosols during pipetting process.
Brand: Greiner Bio-One 740288
Additional Details : Weight : 1.62000kg
Description
- Minimizes danger of unwanted cross-contamination
- Acts as reliable barrier against the transfer or aerosols into shaft of pipette
- Can be used with all standard pipettes
- Maintain tolerance limits stipulated for pipette tips i
Specifications
1000 μL | |
Sterile | |
Irradiation | |
Yes |
Filtered | |
White | |
Anthos™ New Line, Costar™ Onepette™, Eppendorf™ Research™, Thermo Scientific™ Finnpipette™ Digital, Gilson™ Pipetman™, Gilson™ Pipetman™ P200, Human™ Pipettes | |
Polypropylene |