missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Unipur Carbon FT Safety Gloves

Catalog Number 17210461
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Sensitive, anti-static safety gloves feature a thin elastomer coating on the fingertips for precision work with electronics parts.

  • Suited for precision mechanical work in antistatic environments
  • Thin elastomer coating on the fingertips for enhanced grip and tactility
  • Polyamide/carbon liner provides a close fit for increased dexterity
  • Fulfils the requirements of DIN EN 16350:2014
Certifications/Compliance (Europe) DIN EN 16350:2014
Coated Area Fingertips
Applications Industrial
Coating Material Elastomer
Color Gray
Material Polyamide, Carbon Fiber
Size 10
EN388 Standard 2131X
For Use With (Application) Electronics, Precision Assembly
Gender Unisex
Glove Cuff Style Knit
Series Unipur Carbon FT
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'