missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Unidur 6649 Cut Protection Gloves

Catalog Number 17210441
Click to view available options
Size:
7
8
9
This item is not returnable. View return policy
This item is not returnable. View return policy

Lightweight, HPPE gloves feature a polyurethane coating on the palm and fingertips to provide level B cut protection.

  • HPPE fibers provide level B cut protection
  • Polyurethane coating on the palm and fingertips
  • Enhanced grip in dry or slightly damp applications
  • Dirt-resistant exterior is ideal for mechanical applications
Applications Industrial
Coating Material Polyurethane
Color Blue, Gray
Material HPPE, Polyamide, Elastane
Size 7
EN388 Standard 4342B
For Use With (Application) Automotive, Metal Work, and Packaging
Gender Unisex
Glove Cuff Style Knit
Series Unidur 6649
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'