Learn More
Abeba™ UNI6 6751 Shoes
Special lining with silver ions absorbs resperation allowing the foot to breathe. Sanitized™ treated.
Brand:  Abeba™ 6751/44
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 15622098
  • 97.80€ / 1 pair
Estimated Shipment
20-06-2019
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O2 FO SRC
  • Smooth leather
  • Black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • Antistatic
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
Specifications
Specifications
CE EN ISO 20347:2012 O2 FO SRC
Microfiber
44
Polyurethane
Black
Leather
Slip resistant
6751
Videos