Learn More
Abeba™ UNI6 6731 Shoes
Special lining with silver ions absorbs resperation allowing the foot to breathe. Sanitized™ treated.
Brand:  Abeba™ 6731/42
Additional Details : Weight : 0.98000kg
 View more versions of this product
Product Code. 15622068
  • 98.90€ / 1 pair
Estimated Shipment
19-08-2019
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O1 FO SRC
  • Smooth leather, perforated
  • Black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
Specifications
Specifications
CE EN ISO 20347:2012 O1 FO SRC
Leather
Slip resistant
6731
Black
42
Polyurethane