Learn More
Abeba™ UNI6 1784 Shoes
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
Brand:  Abeba™ 1784/46
Additional Details : Weight : 1.12000kg
 View more versions of this product
Product Code. 15627797
  • 88.80€ / 1 pair
Estimated Shipment
20-06-2019
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S2 SRC
  • A-micro
  • Gray/Yellow
  • Steel toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350122)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver Point
  • Instep area with lacing
  • Discontinued model
  • **Size 35 with replaceable acc Wave insole (art. 3576)
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRC
A-micro
Slip resistant
1784
Gray, Yellow
46
Polyurethane
Steel Toe
Videos