missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Ubbelohde ASTM Capillary Viscometers
Ubbelohde ASTM Capillary Viscometers are used to determine kinematic viscosity of transparent Newtonian liquids.
Brand: Fisherbrand™
Additional Details : Weight : 0.80000kg
Description
- Determine viscosities using ASTM and DIN testing methods
- Used to determine kinematic viscosity of transparent Newtonian liquids
- All capillary viscometers are supplied with the calibration certificate
Specifications
1 | |
ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate | |
Glass | |
1 | |
Ubbelohde Astm Viscometer | |
2 to 10 cSt |
0.01 | |
Calibration certificate | |
±0.2% | |
0.25kg | |
15mL |