Learn More
Fisherbrand™ Ubbelohde DIN Capillary Viscometers
Ubbelohde DIN Capillary Viscometers are used to determine kinematic viscosity of transparent Newtonian liquids.
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11846501
Quantity Price
1 166.85€ / Each
10 140.77€ / Each
Estimated Shipment
20-01-2021
Add to basket
Description
Description
  • Determine viscosities using DIN testing method
  •  Used to determine kinematic viscosity of transparent Newtonian liquids
  • All capillary viscometers are supplied with the calibration certificate
Specifications
Specifications
Ic
DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate
Glass
Ic
Ubbelohde Din Viscometer
6 to 30 cSt
0.03
Calibration certificate
±0.2%
0.25kg
15mL
Videos
SDS
Documents
View accessory products