Learn More
Fisherbrand™ Ubbelohde DIN Capillary Viscometers
Ubbelohde DIN Capillary Viscometers are used to determine kinematic viscosity of transparent Newtonian liquids.
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11856501
Quantity Price
1 166.85€ / Each
10 140.77€ / Each
Estimated Shipment
18-01-2021
Add to basket
Description
Description
  • Determine viscosities using DIN testing method
  •  Used to determine kinematic viscosity of transparent Newtonian liquids
  • All capillary viscometers are supplied with the calibration certificate
Specifications
Specifications
Ia
DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate
Glass
Ia
Ubbelohde Din Viscometer
10 to 50 cSt
0.05
Calibration certificate
±0.2%
0.25kg
15mL
Videos
SDS
Documents
View accessory products