Learn More
Fisherbrand™ Ubbelohde DIN Capillary Viscometers
Ubbelohde DIN Capillary Viscometers are used to determine kinematic viscosity of transparent Newtonian liquids.
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11846511
Quantity Price
1 166.85€ / Each
10 140.77€ / Each
Estimated Shipment
21-01-2021
Add to basket
Description
Description
  • Determine viscosities using DIN testing method
  •  Used to determine kinematic viscosity of transparent Newtonian liquids
  • All capillary viscometers are supplied with the calibration certificate
Specifications
Specifications
IVa
DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate
Glass
IVa
Ubbelohde Din Viscometer
10000 to 50000 cSt
50
Calibration certificate
±0.2%
0.25kg
15mL