missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ U-Chem 3300 Chemical Protection Gloves

Catalog Number 17210531
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

NBR-coated chemical protection gloves feature seamless, bamboo-viscose support material that is particularly kind to sensitive skin. Natural fibers help absorb moisture to prevent excessive perspiration.

  • NBR-coated chemical protection gloves with bamboo-fiber supporting material
  • Natural fibers are especially kind to sensitive skin
  • Comfortable to wear for extended periods of time
  • Resistant to a wide variety of chemicals
  • Silicone-free according to imprint test
Applications Laboratory
Coating Material NBR
Color Blue
Size 11
EN388 Standard 2121X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLOPT
For Use With (Application) Chemical Handling, Food Processing, Inspection, Laboratory, Maintenance, and Pharmaceuticals
Gender Unisex
Glove Cuff Style Gauntlet
Liner Material Bamboo, Nylon
Series U-Chem 3300
Length (Metric) 32 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'