missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TrkB [p Tyr816] Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
Specifications
Specifications
| Gene ID (Entrez) | 4915 |
| Species | Rat |
| Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-03499 |
| Gene Alias | BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B |
| Gene Symbol | NTRK2 |
| Quantity | 0.05 mg |
| Research Category | Phospho Specific, Protein Kinase, Signal Transduction |
| Storage Buffer | From PBS. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction